missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, Solstice

Product Code. 15613980
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15613980 2.5 L 2.5 L
15603980 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15613980 Supplier Fluka Supplier No. 334572.5L
  • ONLINE EXCLUSIVE PRICE
    168.00 €
    124.25 € / 2.5 L
    Save 43.75 €
    26% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00002867
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Density 0.966-0.968g/mL at 20°C, 0.965g/mL at 25°C
Explosion Limit 0.245
Flash Point 40°C (104°F)
Ignition Point 548°F
Linear Formula CH3OCH2CH2OH
Quantity 2.5 L
UN Number UN1188
Beilstein 1731074
Formula Weight 76.09g/mol
Vapor Pressure 6.17 mmHg (20°C)
Percent Purity ≥99.5% (GC)
Chemical Name or Material 2-Methoxyethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.