missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, TRC

Product Code. 30132990
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132990 500 mL 500mL
30133634 250 mL 250mL
30011743 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132990 Supplier Toronto Research Chemicals Supplier No. TRCM262435500ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3 H8 O2
Molecular Weight (g/mol) 76.09
Synonym Ethanol, 2-methoxy- (8CI, 9CI, ACI), 1-Hydroxy-2-methoxyethane, 2-Methoxy-1-ethanol, 2-Methoxyethanol, 2-Methoxyethyl alcohol, 3-Oxa-1-butanol, Amsco-Solv EE, Dowanol EM, Ektasolve EM, Ethylene glycol methyl ether, Ethylene glycol monomethyl ether, Glycol methyl ether, Glycol monomethyl ether, Hisolve MC, Methoxyethanol, Methoxyethylene glycol, Methoxyhydroxyethane, Methyl Cellosolve, Methyl glycol, Methyl oxitol, Monoethylene glycol methyl ether, Monomethylglycol, NSC 1258, Poly-Solv EM, β-Methoxyethanol, Ethyleneglycol-monomethyl ether
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Quantity 500 mL
InChI Formula InChI=1S/C3H8O2/c1-5-3-2-4/h4H,2-3H2,1H3
Formula Weight 76.0524
Chemical Name or Material 2-Methoxyethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.