missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethoxymethyl chloride, 94%

Product Code. 11394424
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394424 25 g 25g
11384424 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11394424 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor L01050.14
 Restricted Item
Estimated Shipment: 07-12-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 3970-21-6 | C4H9ClO2 | 124.56 g/mol

2-Methoxyethoxymethyl chloride, is used as selectively cleaved under aprotic conditions in the presence of a wide range of OH-protected reagents. It is used as an OH-protecting reagent. An examples of the target molecule MEM Chloride is the side chain of roxithromycin. It is also used in the protection of the OH groups in serine and threonine during peptide synthesis. Some of the other applications include have the ability to coordinate to metals, which is thought to accelerate the cleavage by Lewis acids. The chelating ability of the MEM ether also makes it useful as a stereodirecting group in organometallic reactions, first noted in the stereo controlled addition of ?-methoxyvinyllithium to a carbonyl in the synthesis of taxusin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxyethoxymethyl chloride, is used as selectively cleaved under aprotic conditions in the presence of a wide range of OH-protected reagents. It is used as an OH-protecting reagent. An examples of the target molecule MEM Chloride is the side chain of roxithromycin. It is also used in the protection of the OH groups in serine and threonine during peptide synthesis. Some of the other applications include have the ability to coordinate to metals, which is thought to accelerate the cleavage by Lewis acids. The chelating ability of the MEM ether also makes it useful as a stereodirecting group in organometallic reactions, first noted in the stereo controlled addition of ɑ-methoxyvinyllithium to a carbonyl in the synthesis of taxusin.

Solubility
It hydrolyzes with water.

Notes
Moisture, Heat, water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 3970-21-6
Molecular Formula C4H9ClO2
Molecular Weight (g/mol) 124.56
MDL Number MFCD00000888
InChI Key BIAAQBNMRITRDV-UHFFFAOYSA-N
Synonym 2-methoxyethoxymethyl chloride, 1-chloromethoxy-2-methoxyethane, ethane, 1-chloromethoxy-2-methoxy, mem chloride, methoxyethoxymethyl chloride, mem-chloride, 1-chloro-2,5-dioxahexane, 2-methoxyethoxy methyl chloride, beta-methoxyethoxymethyl chloride, .beta.-methoxyethoxymethyl chloride
PubChem CID 77590
IUPAC Name 1-(chloromethoxy)-2-methoxyethane
SMILES COCCOCCl

Especificaciones

Density 1.094
Boiling Point 50°C to 52°C (13 mmHg)
Flash Point 54°C (129°F)
Linear Formula CH3O(CH2)2OCH2Cl
Refractive Index 1.427
Quantity 25 g
UN Number UN1992
Beilstein 1900576
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes with water.
Formula Weight 124.57
Percent Purity 94%
Chemical Name or Material 2-Methoxyethoxymethyl chloride
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.