missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl acrylate, 98%, stab. with ca 50-100ppm 4-methoxyphenol

Product Code. 11302707
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302707 50 g 50g
11312707 250 g 250g
11322707 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11302707 Supplier Thermo Scientific Alfa Aesar Supplier No. L11902.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3121-61-7 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3121-61-7
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00048149,MFCD00803685
InChI Key HFCUBKYHMMPGBY-UHFFFAOYSA-N
Synonym 2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca
PubChem CID 18392
IUPAC Name 2-methoxyethyl prop-2-enoate
SMILES COCCOC(=O)C=C

Specifications

Melting Point <-30°C
Density 1.008
Boiling Point 162°C
Flash Point 56°C (133°F)
Odor Ester-like
Linear Formula H2C=CHCO2(CH2)2OCH3
Refractive Index 1.428
Quantity 50 g
UN Number UN2929
Beilstein 1754333
Formula Weight 130.14
Percent Purity 98%
Chemical Name or Material 2-Methoxyethyl acrylate, stabilized with ≈50 to 100ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.