missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl isothiocyanate, 98+%

Product Code. 11393016
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393016 1 g 1g
11303026 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393016 Supplier Thermo Scientific Alfa Aesar Supplier No. L10839.03
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38663-85-3 | C4H7NOS | 117.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38663-85-3
Molecular Formula C4H7NOS
Molecular Weight (g/mol) 117.166
MDL Number MFCD00041126
InChI Key HTSAVXAFEVUJQE-UHFFFAOYSA-N
Synonym 2-methoxyethyl isothiocyanate, ethane, 1-isothiocyanato-2-methoxy, 2-methoxyethylisothiocyanate, 1-isothiocyanato-2-methoxy-ethane, 1-isothiocyanato-2-methoxyethane, 2-methoxyethanisothiocyanate, acmc-1aezw, 2-methoxy ethyl isothiocyanate, #, ethane,1-isothiocyanato-2-methoxy
PubChem CID 123475
IUPAC Name 1-isothiocyanato-2-methoxyethane
SMILES COCCN=C=S

Specifications

Density 1.08
Boiling Point 88°C to 92°C (24 mmHg)
Flash Point 66°C (150°F)
Refractive Index 1.512
Quantity 1 g
UN Number UN2922
Beilstein 773990
Sensitivity Moisture sensitive
Formula Weight 117.17
Percent Purity ≥98%
Chemical Name or Material 2-Methoxyethyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.