missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxynaphthalene, 98%

Product Code. 11463218
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463218 250 g 250g
11453218 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463218 Supplier Thermo Scientific Alfa Aesar Supplier No. A17650.30
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93-04-9 | C11H10O | 158.2 g/mol

2-methoxynaphthalene acylation is used as a model reaction to study the catalytic benefits of delamination. It was also used to study the alkali-metal-mediated manganation (AMMMn) reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-methoxynaphthalene acylation is used as a model reaction to study the catalytic benefits of delamination. It was also used to study the alkali-metal-mediated manganation (AMMMn) reactions.

Solubility
Soluble in benzene, carbon disulfide, diethyl ether. Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-04-9
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.2
MDL Number MFCD00004061
InChI Key LUZDYPLAQQGJEA-UHFFFAOYSA-N
Synonym naphthalene, 2-methoxy, yara yara, nerolin, methyl 2-naphthyl ether, yura yara, 2-naphthyl methyl ether, yara-yara, 2-naphthol methyl ether, nerolin old, beta-naphthyl methyl ether
PubChem CID 7119
IUPAC Name 2-methoxynaphthalene
SMILES COC1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 70°C to 75°C
Boiling Point 272°C to 274°C
Flash Point ca 139°C (282°F)
Quantity 250 g
Beilstein 1859408
Merck Index 14,5997
Solubility Information Soluble in benzene,carbon disulfide,diethyl ether. Insoluble in water.
Formula Weight 158.2
Percent Purity 98%
Chemical Name or Material 2-Methoxynaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.