missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenol, TRC

Product Code. 30011163
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011163 2.5 g 2.5g
30003238 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011163 Supplier Toronto Research Chemicals Supplier No. TRCM2653002.5G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-05-1
Molecular Formula C7 H8 O2
Molecular Weight (g/mol) 124.14
Synonym Guaiacol, 2-Methoxyphenol, Guaifenesin Imp. A (EP), 1-Hydroxy-2-methoxybenzene, 2-Hydroxyanisole, Anastil, Guaiastil, Methylcatechol, NSC 3815, O-Methyl catechol, Pyrocatechol monomethyl ether, Pyroguaiac acid, o-Guaiacol, o-Hydroxyanisole, o-Methoxyphenol
IUPAC Name 2-methoxyphenol
SMILES COc1ccccc1O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8H,1H3
Formula Weight 124.0524
Chemical Name or Material Guaiacol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.