missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypropene, 95%, stab. with ca 0.5% potassium carbonate

Product Code. 11401087
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Product Code. Quantity unitSize
11401087 50 g 50g
11411087 250 g 250g
2 options
This item is not returnable. View return policy

Product Code. 11401087

Brand: Thermo Scientific Alfa Aesar B24083.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 116-11-0 | C4H8O | 72.107 g/mol

It is used for Synthesis of heterocycle compounds and an important intermediate of medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for Synthesis of heterocycle compounds and an important intermediate of medicine.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in a cool, dry place in tightly closed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-11-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00014929
InChI Key YOWQWFMSQCOSBA-UHFFFAOYSA-N
Synonym 2-methoxypropene, isopropenyl methyl ether, 1-propene, 2-methoxy, 2-methoxy-1-propene, methyl isopropenyl ether, ether, isopropenyl methyl, propene, 2-methoxy, unii-15wbg0jt6b, 2-methoxy propene, 15wbg0jt6b
PubChem CID 8300
IUPAC Name 2-methoxyprop-1-ene
SMILES CC(=C)OC

Specifications

Density 0.758
Boiling Point 35°C to 36°C
Flash Point −29°C (−20°F)
Odor Ether-like
Refractive Index 1.3835
Quantity 50 g
UN Number UN1993
Beilstein 1734635
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 72.11
Percent Purity 95%
Chemical Name or Material 2-Methoxypropene, stabilized with ≈0.5% potassium carbonate
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.