missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypropene, TRC

Product Code. 30481447
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
Product Code. Quantity unitSize
30481447 250 g 250g
1 options
This item is not returnable. View return policy

Product Code. 30481447

Brand: Toronto Research Chemicals TRCM265055250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methoxypropene
CAS 116-11-0
Molecular Formula C4H8O
Quantity 250 g
Synonym 2-Methoxyprop-1-ene, 2-Methoxypropene, Isopropenyl Methyl Ether, Methyl 1-Methylvinyl Ether, Methyl Isopropenyl Ether
InChI Formula InChI=1S/C4H8O/c1-4(2)5-3/h1H2,2-3H3
SMILES COC(=C)C
IUPAC Name 2-methoxyprop-1-ene
Molecular Weight (g/mol) 72.11
Formula Weight 72.0575
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.