missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3(2H,4H)-isoquinolinedione, TRC

Product Code. 30474092
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474092 10 mg 10mg
30482797 5 mg 5mg
30465598 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474092 Supplier Toronto Research Chemicals Supplier No. TRCM22349310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4494-53-5
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.18
Synonym 2-Methylisoquinoline-1,3(2H,4H)-dione
IUPAC Name 2-methyl-4H-isoquinoline-1,3-dione
SMILES CN1C(=O)Cc2ccccc2C1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H9NO2/c1-11-9(12)6-7-4-2-3-5-8(7)10(11)13/h2-5H,6H2,1H3
Formula Weight 175.0633
Chemical Name or Material 2-Methyl-1,3(2H,4H)-isoquinolinedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.