missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-benzothiazol-6-ylamine, 97%, Thermo Scientific™

Product Code. 10431514
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10431514 250 mg 250mg
10708835 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10431514 Supplier Thermo Scientific Maybridge Supplier No. CC54714CB
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2941-62-0
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.226
InChI Key HFUJOSYKJMNSFQ-UHFFFAOYSA-N
Synonym 2-methylbenzo d thiazol-6-amine, 6-amino-2-methylbenzothiazole, 2-methyl-benzothiazol-6-ylamine, 6-benzothiazolamine, 2-methyl, 2-methyl-1,3-benzothiazol-6-ylamine, 6-amino-2-methyl-benzothiazole, 2-methylbenzothiazol-6-amine, 2-methylbenzothiazole-6-ylamine, pubchem20725, acmc-1cctf
PubChem CID 762831
IUPAC Name 2-methyl-1,3-benzothiazol-6-amine
SMILES CC1=NC2=C(S1)C=C(C=C2)N

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 164.23
Percent Purity 97%
Chemical Name or Material 2-Methyl-1, 3-benzothiazol-6-ylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.