missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-thiazole-4-carboximidamide hydrochloride, 97%, Thermo Scientific™

Product Code. 10752323
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10752323 1 g Amber Glass Bottle 1g
10544994 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10752323 Supplier Thermo Scientific Maybridge Supplier No. SPB03559DA
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 18876-82-9
Molecular Formula C5H8ClN3S
Molecular Weight (g/mol) 177.65
MDL Number MFCD00178765
InChI Key YYLYRAUCGUIGIE-UHFFFAOYSA-N
Synonym 2-methyl-1,3-thiazole-4-carboximidamide hydrochloride, 2-methylthiazole-4-carboxamidine hydrochloride, 2-methyl-thiazole-4-carboxamidine hydrochloride, 2-methylthiazole-4-carboximidamide hydrochloride, 2-methyl-1,3-thiazole-4-carboxamidine, chloride, 4-thiazolecarboximidamide,2-methyl-,hydrochloride 1:1
PubChem CID 2799402
IUPAC Name 2-methyl-1,3-thiazole-4-carboximidamide;hydrochloride
SMILES CC1=NC(=CS1)C(=N)N.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 177.66
Percent Purity 97%
Chemical Name or Material 2-Methyl-1, 3-thiazole-4-carboximidamide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.