missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10394702
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10394702 250 mg 250mg
10572662 1 g 1g
10321665 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10394702 Supplier Thermo Scientific Maybridge Supplier No. CC05701CB
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 35272-15-2
Molecular Formula C5H5NO2S
Molecular Weight (g/mol) 143.16
MDL Number MFCD03407332
InChI Key ZHDRDZMTEOIWSX-UHFFFAOYSA-N
Synonym 2-methylthiazole-4-carboxylic acid, 2-methyl-4-thiazolecarboxylic acid, 4-thiazolecarboxylic acid, 2-methyl, 2-methyl-thiazole-4-carboxylic acid, 2-methyl-4-thiazolecarboxylicacid, 2-methyl-thiazole4-carboxylic acid, 4-thiazolecarboxylicacid, 2-methyl, 2-methyl-1,3-thiazole-4-carboxylicacid
PubChem CID 284728
IUPAC Name 2-methyl-1,3-thiazole-4-carboxylic acid
SMILES CC1=NC(=CS1)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 143.17
Percent Purity 97%
Chemical Name or Material 2-Methyl-1, 3-thiazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.