missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,4-butanediol, TRC

Product Code. 30468496
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30468496 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468496 Supplier Toronto Research Chemicals Supplier No. TRCM22249550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 2-Methyl-1,4-butanediol
CAS 2938-98-9
Molecular Formula C5H12O2
Quantity 50 mg
InChI Formula InChI=1S/C5H12O2/c1-5(4-7)2-3-6/h5-7H,2-4H2,1H3
SMILES C(CCO)(CO)C
IUPAC Name 2-methylbutane-1,4-diol
Molecular Weight (g/mol) 104.15
Formula Weight 104.0837
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.