missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1,4-naphthoquinone-d8, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Vitamin K3-d8 |
| CAS | 478171-80-1 |
| Molecular Formula | C11 2H8 O2 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C11H8O2/c1-7-6-10(12)8-4-2-3-5-9(8)11(7)13/h2-6 H,1H3/i1D3,2 D,3 D,4 D,5 D,6 D |
| SMILES | [2 H]C1=C(C(=O)c2c([2 H])c([2 H])c([2 H])c([2 H])c2C1=O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,5,6,7,8-pentadeuterio-3-(trideuteriomethyl)naphthalene-1,4-dione |
| Molecular Weight (g/mol) | 180.23 |
| Formula Weight | 180.1026 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction