missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-buten-3-yne, 97%

Product Code. 11342386
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342386 25 g 25g
11332386 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342386 Supplier Thermo Scientific Alfa Aesar Supplier No. L06111.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78-80-8 | C5H6 | 66.103 g/mol

2-Methyl-1-buten-3-yne is a reagent used in analytical studies to determine the aroma compounds present in various things by using pyrolysis gas chromatography mass spectrometry techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1-buten-3-yne is a reagent used in analytical studies to determine the aroma compounds present in various things by using pyrolysis gas chromatography mass spectrometry techniques.

Solubility
Not miscible in water.

Notes
Store at 4°C. Incompatible with heavy metal powders, heat. Protect from heat and direct sunlight. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-80-8
Molecular Formula C5H6
Molecular Weight (g/mol) 66.103
MDL Number MFCD00008599
InChI Key BOFLDKIFLIFLJA-UHFFFAOYSA-N
Synonym isopropenylacetylene, 2-methyl-1-buten-3-yne, valylene, 1-buten-3-yne, 2-methyl, 3-methyl-3-buten-1-yne, 2-methylbutenyne, isopropenyl acetylene, acmc-20anrd, 2-methyl-1-butenyne, 2-methyl-l-buten-3-yne
PubChem CID 62323
IUPAC Name 2-methylbut-1-en-3-yne
SMILES CC(=C)C#C

Specifications

Melting Point -113°C
Density 0.694
Boiling Point 32°C to 33°C
Flash Point −45°C (−49°F)
Odor Pungent
Refractive Index 1.418
Quantity 25 g
UN Number UN3295
Beilstein 969175
Solubility Information Not miscible in water.
Formula Weight 66.1
Percent Purity 97%
Chemical Name or Material 2-Methyl-1-buten-3-yne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.