missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-propanol, CHROMASOLV™, for HPLC, 99.5%, Honeywell Riedel-de Haen

Product Code. 15692840
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15692840 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15692840 Supplier Honeywell-Chromasolv Supplier No. 2704661L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CHROMASOLV™, for HPLC, 99.5%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -108°C (literature)
Absorbance max. 0.50 at 230 nm, max. 1.0 at 219 nm, max. 0.01 at 400 nm, max. 0.01 at 330 nm, max. 0.03 at 260 nm, max. 0.05 at 250 nm, max. 0.10 at 245 nm
CAS 78-83-1
CAS Max % 100.0000%
Density 0.803 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point 28°C
Packaging Glass Bottle
Molecular Formula C4H10O
Linear Formula (CH3)2CHCH2OH
MDL Number MFCD00004740
UN Number UN1212
Beilstein 1730878
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
Boiling Point 108°C
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
SMILES CC(C)CO
IUPAC Name 2-methylpropan-1-ol
Molecular Weight (g/mol) 74.123
PubChem CID 6560
ChEBI CHEBI:46645
Formula Weight 74.12
Percent Purity >99.45%
Grade HPLC
Chemical Name or Material 2-Methyl-1-propanol, CHROMASOLV™, for HPLC, 99.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.