missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-pyrroline, 98%, Thermo Scientific Chemicals

Product Code. 10241323 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10241323 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10241323 Supplier Thermo Scientific Acros Supplier No. 135070250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 872-32-2 | C5H10N | 84.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-32-2
Molecular Formula C5H10N
Molecular Weight (g/mol) 84.14
MDL Number MFCD00128806
InChI Key OZLJZWIVBYLSOK-UHFFFAOYSA-N
Synonym 2-methyl-1-pyrroline, 3,4-dihydro-5-methyl-2h-pyrrole, 1pyrroline2methyl, 2h-pyrrole, 3,4-dihydro-5-methyl, pubchem9412
PubChem CID 70103
ChEBI CHEBI:78856
SMILES C[C+]1CCCN1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Beige to White
Density 0.8780g/mL
Boiling Point 104.0°C to 105.0°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420
Quantity 25 mL
Specific Gravity 0.878
Solubility Information Solubility in water: soluble.
IUPAC Name 5-methyl-3,4-dihydro-2H-pyrrole
Formula Weight 83.13
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 2-Methyl-1-pyrroline, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.