missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-butanol, 99+%, extra pure, ACROS Organics™

Product Code. 10471641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
Unit Size:
5mL
Product Code. Quantity Packaging unitSize
10471641 5 mL Glass bottle 5mL
1 options
This item is not returnable. View return policy

Product Code. 10471641

Brand: Thermo Scientific Acros 220230050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 75-85-4
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C5H12O
Linear Formula C2H5C(CH3)2OH
Refractive Index 1.4040 to 1.4060 (20°C, 589nm)
MDL Number MFCD00004478
Quantity 5 mL
Beilstein 01, 388
Residue after Evaporation 0.001% max.
Merck Index 15, 7253
Synonym 2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol
InChI Key MSXVEPNJUHWQHW-UHFFFAOYSA-N
SMILES CCC(C)(C)O
IUPAC Name 2-methylbutan-2-ol
Molecular Weight (g/mol) 88.15
PubChem CID 6405
Formula Weight 88.15
Percent Purity ≥99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-butanol, p.a.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.