missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-cyclopenten-1-one, 97%

Product Code. 10462821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10462821 5 g Glass bottle 5g
10056253 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10462821 Supplier Thermo Scientific Acros Supplier No. 270360050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1120-73-6 | C6H8O | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-73-6
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD00012275
InChI Key ZSBWUNDRDHVNJL-UHFFFAOYSA-N
Synonym 2-methylcyclopent-2-enone, 2-methyl-2-cyclopenten-1-one, 2-cyclopenten-1-one, 2-methyl, 2-methyl-2-cyclopentenone, cyclopentenone, 2-methyl, cyclopentenone, methyl, methyl-cyclopentenone, acmc-20af73, 2-cyclopenten-1-one, methyl, 2-cyclopenten-1-one,2-methyl
PubChem CID 14266
IUPAC Name 2-methylcyclopent-2-en-1-one
SMILES CC1=CCCC1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 0.9790g/mL
Boiling Point 158.0°C to 161.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3C5H5(0)
Refractive Index 1.4780 to 1.4800
Quantity 5 g
Beilstein 07, 52
Specific Gravity 0.979
Formula Weight 96.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-cyclopenten-1-one, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-2-cyclopenten-1-one
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.