missing translation for 'onlineSavingsMsg'
Learn More

2-methyl-2-(morpholin-4-yl)propanoic Acid hydrochloride, TRC

Product Code. 30476531
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476531 100 mg 100mg
30481875 50 mg 50mg
30472995 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30476531 Supplier Toronto Research Chemicals Supplier No. TRCM357943100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375397-37-8
Molecular Formula C8H15NO3.HCl
Molecular Weight (g/mol) 209.67
Synonym 4-Morpholineacetic acid, α,α-dimethyl-, hydrochloride (1:1) (ACI), 4-Morpholineacetic acid, α,α-dimethyl-, hydrochloride (9CI), 2-Methyl-2-(morpholin-4-yl)propanoic acid hydrochloride, α,α-Dimethyl-4-morpholineacetic acid hydrochloride
IUPAC Name 2-methyl-2-morpholin-4-ylpropanoic acid;hydrochloride
SMILES Cl.CC(C)(N1CCOCC1)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H15NO3.ClH/c1-8(2,7(10)11)9-3-5-12-6-4-9;/h3-6H2,1-2H3,(H,10,11);1H
Formula Weight 209.0819
Chemical Name or Material 2-methyl-2-(morpholin-4-yl)propanoic Acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.