missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-oxazoline, 99%, Thermo Scientific Chemicals

Catalog Number 10427190
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1120-64-5
C4H7NO
85.11
MFCD00005298
GUXJXWKCUUWCLX-UHFFFAOYSA-N
2-methyl-2-oxazoline, 2-methyloxazoline, 2-methyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-2-methyl, 2-methyl-2-oxazolin, 2-methyl-1,3-oxazoline, 2-oxazoline, 2-methyl, 4,5-dihydro-2-methyl-1,3-oxazole, poly 2-methyl-2-oxazoline, methyl oxazoline
70713
CHEBI:53614
2-methyl-4,5-dihydro-1,3-oxazole
CC1=NCCO1
This item is not returnable. View return policy
1120-64-5
C4H7NO
85.11
MFCD00005298
GUXJXWKCUUWCLX-UHFFFAOYSA-N
2-methyl-2-oxazoline, 2-methyloxazoline, 2-methyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-2-methyl, 2-methyl-2-oxazolin, 2-methyl-1,3-oxazoline, 2-oxazoline, 2-methyl, 4,5-dihydro-2-methyl-1,3-oxazole, poly 2-methyl-2-oxazoline, methyl oxazoline
70713
CHEBI:53614
2-methyl-4,5-dihydro-1,3-oxazole
CC1=NCCO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 109.4°C to 110.4°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4330 to 1.4350
Quantity 100 g
Beilstein 27, 13
Specific Gravity 1
Formula Weight 85.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-oxazoline, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-2-oxazoline
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'