missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-pentenal, 97%

Product Code. 10622233 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10622233 25 g Glass bottle 25g
10380474 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10622233 Supplier Thermo Scientific Acros Supplier No. 359960250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 623-36-9 | C6H10O | 98.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-36-9
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00006978
InChI Key IDEYZABHVQLHAF-GQCTYLIASA-N
Synonym 2-methyl-2-pentenal, 2-pentenal, 2-methyl, e-2-methyl-2-pentenal, 2-methyl-3-ethylacrolein, 2-methyl-2-penten-1-al, 2-methyl-2-pentene-1-al, 2,4-dimethylcrotonaldehyde, trans-2-methyl-2-pentenal, e-2-methylpent-2-enal, unii-98qag1u530
PubChem CID 5319754
IUPAC Name (E)-2-methylpent-2-enal
SMILES CCC=C(C)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 134.0°C to 136.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH=C(CH3)CHO
Refractive Index 1.4470 to 1.4550
Quantity 25 g
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble
Formula Weight 98.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-pentenal
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-2-pentenal
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.