missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-pentenal, (E)+(Z), 97%

Product Code. 11462358
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462358 25 g 25g
11472358 100 g 100g
11482358 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462358 Supplier Thermo Scientific Alfa Aesar Supplier No. A15729.14
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 623-36-9 | C6H10O | 98.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-36-9
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00006978
InChI Key IDEYZABHVQLHAF-GQCTYLIASA-N
Synonym 2-methyl-2-pentenal, 2-pentenal, 2-methyl, e-2-methyl-2-pentenal, 2-methyl-3-ethylacrolein, 2-methyl-2-penten-1-al, 2-methyl-2-pentene-1-al, 2,4-dimethylcrotonaldehyde, trans-2-methyl-2-pentenal, e-2-methylpent-2-enal, unii-98qag1u530
PubChem CID 5319754
IUPAC Name (E)-2-methylpent-2-enal
SMILES CCC=C(C)C=O

Specifications

Melting Point -90°C
Density 0.86
Boiling Point 134°C to 136°C
Flash Point 31°C (87°F)
Odor Pleasant
Refractive Index 1.4495
Quantity 25 g
UN Number UN1988
Beilstein 506124
Sensitivity Air Sensitive
Formula Weight 98.15
Percent Purity 97%
Chemical Name or Material 2-Methyl-2-pentenal, (E)+(Z)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.