missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-thiazoline, 97%

Product Code. 11384554
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384554 25 g 25g
11394554 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384554 Supplier Thermo Scientific Alfa Aesar Supplier No. L01329.14
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2346-00-1 | C4H7NS | 101.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2346-00-1
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.17
MDL Number MFCD00005314
InChI Key JUIQOABNSLTJSW-UHFFFAOYSA-N
Synonym 2-methyl-2-thiazoline, 2-methylthiazoline, thiazole, 4,5-dihydro-2-methyl, 2-methyl-4,5-dihydrothiazole, 4,5-dihydro-2-methylthiazole, methyl-2-thiazoline, 2-thiazoline, 2-methyl, unii-8z6ua8v7wn, methyl-2 delta-2 thiazoline, methyl-2 delta-2 thiazoline french
PubChem CID 16867
IUPAC Name 2-methyl-4,5-dihydro-1,3-thiazole
SMILES CC1=NCCS1

Specifications

Melting Point -101°C
Boiling Point 144°C to 145°C
Flash Point 37°C (98°F)
Quantity 25 g
UN Number UN1993
Beilstein 104274
Formula Weight 101.17
Percent Purity 97%
Chemical Name or Material 2-Methyl-2-thiazoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.