missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-buten-2-ol, 97%

Product Code. 10656172 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10656172 250 mL Glass bottle 250mL
10153210 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10656172 Supplier Thermo Scientific Acros Supplier No. 159672500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 115-18-4 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-18-4
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00004470
InChI Key HNVRRHSXBLFLIG-UHFFFAOYSA-N
Synonym 2-methyl-3-buten-2-ol, 1,1-dimethylallyl alcohol, methylbutenol, 3-buten-2-ol, 2-methyl, 3-hydroxy-3-methyl-1-butene, dimethylvinylcarbinol, dimethylvinylmethanol, vinyldimethylcarbinol, isoprenyl alcohol, 3-methyl-1-buten-3-ol
PubChem CID 8257
IUPAC Name 2-methylbut-3-en-2-ol
SMILES CC(C)(C=C)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -43.0°C
Color Colorless to Yellow
Density 0.8200g/mL
Boiling Point 98.0°C to 99.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHC(CH3)2OH
Refractive Index 1.4160 to 1.4180
Quantity 250 mL
Beilstein 01, 444
Specific Gravity 0.82
Solubility Information Solubility in water: 100g/L (20 c)
Formula Weight 86.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-buten-2-ol, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-3-buten-2-ol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.