missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-furoic acid, 98%

Product Code. 11491427
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491427 25 g 25g
11481427 5 g 5g
11401437 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491427 Supplier Thermo Scientific Alfa Aesar Supplier No. B24928.14
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6947-94-0 | C6H6O3 | 126.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6947-94-0
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00092314
InChI Key CFGQZVOVFIZRMN-UHFFFAOYSA-N
PubChem CID 244756
IUPAC Name 2-methylfuran-3-carboxylic acid
SMILES CC1=C(C=CO1)C(=O)O

Specifications

Melting Point 97°C to 99°C
Quantity 25 g
Beilstein 112229
Synonym 2-methyl-3-furoic acid, 2-methyl-3-furancarboxylic acid, 3-furancarboxylic acid, 2-methyl, 2-methyl-furan-3-carboxylic acid, 3-furancarboxylic acid,2-methyl, zlchem 1156, acmc-1b2rk, 2-methyl-3furan carboxylic acid, 2-methyl-3-furan carboxylic acid
Formula Weight 126.11
Percent Purity 98%
Chemical Name or Material 2-Methyl-3-furoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.