missing translation for 'onlineSavingsMsg'
Learn More

(2-Methyl-3-furyl)methanol, 97%, Thermo Scientific™

Product Code. 10565354
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10565354 1 g 1g
10255694 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10565354 Supplier Thermo Scientific Maybridge Supplier No. CD10514DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 5554-99-4
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00052572
InChI Key FTYOGLDPNBZSQC-UHFFFAOYSA-N
PubChem CID 2777165
IUPAC Name (2-methylfuran-3-yl)methanol
SMILES CC1=C(C=CO1)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Synonym 2-methylfuran-3-yl methanol, 2-methyl-3-furyl methanol, 2-methyl-3-furanmethanol, 3-hydroxymethyl-2-methylfuran, 3-furanmethanol, 2-methyl, 2-methyl-furan-3-yl-methanol, 3-furanmethanol,2-methyl, 2-methylfur-3-yl methanol, 2-methyl-3-furyl methan-1-ol
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material (2-Methyl-3-furyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.