missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-(4-morpholinyl)benzenamine, 97%

Product Code. 10893021 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10893021 1 g Glass bottle 1g
10678360 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Código de producto. 10893021 Proveedor Thermo Scientific Acros N.º de proveedor 432460010
 Restricted Item
Estimated Shipment: 19-11-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 581-00-0 | C11H16N2O | 192.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 581-00-0
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.26
MDL Number MFCD10686817
InChI Key ZGJUJDQANIYVAL-UHFFFAOYSA-N
Synonym 2-methyl-4-morpholin-4-yl aniline, 2-methyl-4-morpholinoaniline, 2-methyl-4-4-morpholinyl aniline, 4-p-amino-m-tolyl morpholine, 4-4-morpholinyl-2-methylaniline, 2-methyl4-4-morpholinyl benzenamine, n-4-amino-3-methylphenyl morpholine, 2-methyl-4-4-morpholinyl benzenamine, 2-methyl-4-morpholin-4-yl-phenylamine, benzenamine, 2-methyl-4-4-morpholinyl
PubChem CID 21955000
IUPAC Name 2-methyl-4-morpholin-4-ylaniline
SMILES CC1=C(C=CC(=C1)N2CCOCC2)N

Especificaciones

CAS Min % 96.0
CAS Max % 100.0
Melting Point 91°C to 92°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 192.26
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-4-(4-morpholinyl)benzenamine
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 2-Methyl-4-(4-morpholinyl)benzenamine
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.