missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4,5-bis(methylsulfonyl)-benzoic Acid Methyl Ester, TRC

Product Code. 30478041
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478041 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478041 Supplier Toronto Research Chemicals Supplier No. TRCM292480100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methyl-4,5-bis(methylsulfonyl)-benzoic Acid Methyl Ester
CAS 373357-96-1
Molecular Formula C37 H30 O10
Quantity 100 mg
InChI Formula InChI=1S/C37H30O10/c1-22-19-30(38)43-28-20-26(17-18-27(22)28)42-37-33(46-35(40)24-13-7-3-8-14-24)32(45-34(39)23-11-5-2-6-12-23)31-29(44-37)21-41-36(47-31)25-15-9-4-10-16-25/h2-20,29,31-33,36-37H,21H2,1H3/t29?,31-,32?,33?,36?,37+/m0/s1
SMILES CC1=CC(=O)Oc2cc(O[C@@H]3OC4COC(O[C@@H]4C(OC(=O)c5ccccc5)C3OC(=O)c6ccccc6)c7ccccc7)ccc12
IUPAC Name [(6S,8aS)-7-benzoyloxy-6-(4-methyl-2-oxochromen-7-yl)oxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-8-yl] benzoate
Molecular Weight (g/mol) 634.628
Formula Weight 634.184
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.