missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-n-propyl-1,3-oxathiane, cis + trans, 99%, Thermo Scientific™

Product Code. 11475237
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475237 25 g 25g
11465237 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475237 Supplier Thermo Scientific Alfa Aesar Supplier No. B22630.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67715-80-4
Molecular Formula C8H16OS
Molecular Weight (g/mol) 160.275
MDL Number MFCD00673203
InChI Key GKGOLPMYJJXRGD-UHFFFAOYSA-N
Synonym 1,3-oxathiane, 2-methyl-4-propyl, 2-methyl-4-propyl-1,3-oxathiane, mixture of cis and trans, 1,3-oxathiane, 2-methyl-4-propyl-, 2r,4r-rel, 1,3-oxathiane, 2-methyl-4-propyl-, 2r,4s-rel, tropathiane, z-2-methyl-4-propyl-1,3-oxathiane, 2-tropical oxathiane, e-galbanum oxathiane, z-galbanum oxathiane, dsstox_cid_27704
PubChem CID 101010
ChEBI CHEBI:87335
IUPAC Name 2-methyl-4-propyl-1,3-oxathiane
SMILES CCCC1CCOC(S1)C

Specifications

Boiling Point 218°C to 220°C
Quantity 25 g
UN Number UN1993
Formula Weight 160.27
Percent Purity 99%
Chemical Name or Material 2-Methyl-4-n-propyl-1,3-oxathiane, cis + trans
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.