missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-nitroaniline, TRC

Product Code. 30481108
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481108 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481108 Supplier Toronto Research Chemicals Supplier No. TRCM323545250G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-5-nitrotoluene (2-Methyl-4-nitroaniline)
CAS 99-52-5
Molecular Formula C7 H8 N2 O2
Quantity 250 g
Synonym o-Toluidine','4-nitro- (8CI), 2-Methyl-4-nitrobenzenamine, 1-Amino-2-methyl-4-nitrobenzene, 2-Amino-1-methyl-5-nitrobenzene, 2-Amino-5-nitrotoluene, 2-Methyl-4-nitrophenylamine, 2-Methyl-p-nitroaniline, 3-Methyl-4-aminonitrobenzene, 4-Nitro-2-methylaniline, 4-Nitro-2-toluidine, 4-Nitro-6-methylaniline, 4-Nitro-o-toluidine, 5-Nitro-2-aminotoluene, Ansibases Red RL, Azoene Fast Red GL Base, Azogene Fast Red NRL Salt, Azogene Fast Red RL, Daito Red Base RL, Devol Red RL, Devol Red Salt E, Diabase Red RL, Fast Red 5NT, Fast Red 5NT Salt, Fast Red Base RL, Fast Red RL Base, Fast Red Salt RL, Hiltonil Fast Red RL Base, Hiltosal Fast Red RL Salt, Kako Red RL Base, Kayaku Red RL Base, MNA, MNA (optical crystal), Meisei Fast Red RL Base, Mitsui Red RL Base, NSC 2075, NSC 5508, Naphtoelan Red RL Base, Red Base Ciba X, Red Base Irga X, Red Base NRL, Red RL Base, Red base RL, Sanyo Fast Red RL Base, Sanyo Fast Red Salt RL, Spectrolene Red RL, Symulon Red RL Base, Tulabase Fast Red RL, Yamada Fast Red RL Base
InChI Formula InChI=1S/C7H8N2O2/c1-5-4-6(9(10)11)2-3-7(5)8/h2-4H,8H2,1H3
SMILES Cc1cc(ccc1N)[N+](=O)[O-]
IUPAC Name 2-methyl-4-nitroaniline
Molecular Weight (g/mol) 152.15
Formula Weight 152.0586
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.