missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-oxo-4-phenylbutyric Acid, TRC

Product Code. 30471549
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471549 500 mg 500mg
30470924 100 mg 100mg
30480730 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30471549 Supplier Toronto Research Chemicals Supplier No. TRCM327155500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1771-65-9
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
Synonym 2-Methyl-4-oxo-4-phenylbutanoic Acid
IUPAC Name 2-methyl-4-oxo-4-phenylbutanoic acid
SMILES O=C(C(CC(C1=CC=CC=C1)=O)C)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H12O3/c1-8(11(13)14)7-10(12)9-5-3-2-4-6-9/h2-6,8H,7H2,1H3,(H,13,14)
Formula Weight 192.08
Chemical Name or Material 2-Methyl-4-oxo-4-phenylbutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.