missing translation for 'onlineSavingsMsg'
Learn More

(2-Methyl-4-phenyl-1,3-thiazol-5-yl)methanol, 97%, Thermo Scientific™

Product Code. p-3484471
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11330617 250 mg Amber Glass Bottle 250mg
10604595 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11330617 Supplier Thermo Scientific Maybridge Supplier No. CC42209CB
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 857284-12-9
Molecular Formula C11H11NOS
Molecular Weight (g/mol) 205.275
MDL Number MFCD08690247
InChI Key RBRYERMYEBFVAB-UHFFFAOYSA-N
Synonym 2-methyl-4-phenyl-1,3-thiazol-5-yl methanol, 5-thiazolemethanol,2-methyl-4-phenyl, 2-methyl-4-phenylthiazol-5-yl methanol, 2-methyl-4-phenyl-1,3-thiazol-5-yl methan-1-ol
PubChem CID 18525756
IUPAC Name (2-methyl-4-phenyl-1,3-thiazol-5-yl)methanol
SMILES CC1=NC(=C(S1)CO)C2=CC=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 205.28
Percent Purity 97%
Chemical Name or Material (2-Methyl-4-phenyl-1, 3-thiazol-5-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.