missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-phenyl-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 11320617
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11320617 250 mg Amber Glass Bottle 250mg
10071884 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11320617 Supplier Thermo Scientific Maybridge Supplier No. CC42201CB
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 32002-72-5
Molecular Formula C11H8NO2S
Molecular Weight (g/mol) 218.25
MDL Number MFCD07348569
InChI Key APXSARIYDLVPAY-UHFFFAOYSA-M
Synonym 2-methyl-4-phenylthiazole-5-carboxylic acid, 2-methy-4-phenylthiazol-5-ylcarboxylic acid, 2-methyl-4-phenyl-thiazole-5-carboxylic acid, 2-methyl-4-phenylthiazol-5-ylcarboxylic acid, 5-thiazolecarboxylicacid, 2-methyl-4-phenyl, 5-thiazolecarboxylic acid, 2-methyl-4-phenyl
PubChem CID 7131132
IUPAC Name 2-methyl-4-phenyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=NC(=C(S1)C([O-])=O)C1=CC=CC=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 219.26
Percent Purity 97%
Chemical Name or Material 2-Methyl-4-phenyl-1, 3-thiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.