missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-phenyl-5-pyrimidinecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10202832
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10202832 250 mg 250mg
10354662 1 g 1g
10782197 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10202832 Supplier Thermo Scientific Maybridge Supplier No. CC20101CB
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

Identificatori chimici

CAS 127958-10-5
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.224
MDL Number MFCD02682072
InChI Key XKJAQESOTNACHT-UHFFFAOYSA-N
PubChem CID 2776510
IUPAC Name 2-methyl-4-phenylpyrimidine-5-carboxylic acid
SMILES CC1=NC=C(C(=N1)C2=CC=CC=C2)C(=O)O

Specifica

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 2-methyl-4-phenyl-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylicacid, 2-methyl-4-phenyl, 5-pyrimidinecarboxylic acid, 2-methyl-4-phenyl, acmc-1c0qx, 2-methyl-4-phenylpyrimidin-5-carboxylic acid, 5-pyrimidinecarboxylicacid,2-methyl-4-phenyl, 2-methyl-4-phenyl-pyrimidine-5-carboxylic acid
Formula Weight 214.22
Percent Purity 97%
Chemical Name or Material 2-Methyl-4-phenyl-5-pyrimidinecarboxylic acid
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.