missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-phenyl-5-pyrimidinecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10354662
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10354662 1 g 1g
10202832 250 mg 250mg
10782197 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10354662 Supplier Thermo Scientific Maybridge Supplier No. CC20101DA
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 127958-10-5
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.224
MDL Number MFCD02682072
InChI Key XKJAQESOTNACHT-UHFFFAOYSA-N
PubChem CID 2776510
IUPAC Name 2-methyl-4-phenylpyrimidine-5-carboxylic acid
SMILES CC1=NC=C(C(=N1)C2=CC=CC=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 2-methyl-4-phenyl-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylicacid, 2-methyl-4-phenyl, 5-pyrimidinecarboxylic acid, 2-methyl-4-phenyl, acmc-1c0qx, 2-methyl-4-phenylpyrimidin-5-carboxylic acid, 5-pyrimidinecarboxylicacid,2-methyl-4-phenyl, 2-methyl-4-phenyl-pyrimidine-5-carboxylic acid
Formula Weight 214.22
Percent Purity 97%
Chemical Name or Material 2-Methyl-4-phenyl-5-pyrimidinecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.