missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-piperazinoquinoline, 97%, Thermo Scientific™

Product Code. 10354152
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10354152 1 g 1g
10344152 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10354152 Supplier Thermo Scientific Maybridge Supplier No. RH02136DA
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 82241-22-3
Molecular Formula C14H17N3
Molecular Weight (g/mol) 227.31
MDL Number MFCD01935745
InChI Key XYBLCORUTWKJOI-UHFFFAOYSA-N
Synonym 2-methyl-4-piperazinoquinoline, 2-methyl-4-piperazin-1-yl quinoline, quinoline, 2-methyl-4-1-piperazinyl, 2-methyl-4-quinolyl piperazine, 2-methyl-4-piperazin-1-yl-quinoline, maybridge1_006771, chemdivam_001084, cbmicro_045139, cambridge id 6095001, tos-bb-0571
PubChem CID 702318
SMILES CC1=CC(N2CCNCC2)=C2C=CC=CC2=N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name 2-methyl-4-(piperazin-1-yl)quinoline
Formula Weight 227.31
Percent Purity 97%
Chemical Name or Material 2-Methyl-4-piperazinoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.