missing translation for 'onlineSavingsMsg'
Learn More

2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10624965
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10624965 250 mg Amber Glass Bottle 250mg
10091474 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10624965 Supplier Thermo Scientific Maybridge Supplier No. CC76304CB
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1034566-13-6
Molecular Formula C6H4F3NOS
Molecular Weight (g/mol) 195.159
MDL Number MFCD12198128
InChI Key FBQFBBGXQZDOQV-UHFFFAOYSA-N
Synonym 2-methyl-4-trifluoromethyl-1,3-thiazole-5-carbaldehyde, 5-formyl-2-methyl-4-trifluoromethyl-1,3-thiazole
PubChem CID 43811060
IUPAC Name 2-methyl-4-(trifluoromethyl)-1,3-thiazole-5-carbaldehyde
SMILES CC1=NC(=C(S1)C=O)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 195.16
Percent Purity 97%
Chemical Name or Material 2-Methyl-4-(trifluoromethyl)-1, 3-thiazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.