missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-trimethylsilyl-1-buten-3-yne, 97%

Product Code. 11329688
Change view
Click to view available options
Quantity:
2 g
50 g
Unit Size:
2g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329688 2 g 2g
11309688 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329688 Supplier Thermo Scientific Alfa Aesar Supplier No. 044189.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18387-60-5 | C8H14Si | 138.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18387-60-5
Molecular Formula C8H14Si
Molecular Weight (g/mol) 138.285
MDL Number MFCD00190206
InChI Key HRGBALJHGYAWBL-UHFFFAOYSA-N
Synonym 2-methyl-4-trimethylsilyl-1-buten-3-yne, 2-methyl-4-trimethylsilylbut-1-en-3-yne, acmc-1cdn8, trimethylsilylisopropenylacetylene, trimethyl 3-methylbut-3-en-1-ynyl silane, 3-methyl-3-butene-1-ynyl trimethylsilane, trimethyl 3-methyl-3-buten-1-ynyl silane #, trimethyl 3-methylbut-3-en-1-yn-1-yl silane, silane,trimethyl 3-methyl-3-buten-1-yn-1-yl, 2-methyl-4-trimethylsilyl-1-buten-3-yne,
PubChem CID 579646
IUPAC Name trimethyl(3-methylbut-3-en-1-ynyl)silane
SMILES CC(=C)C#C[Si](C)(C)C

Specifications

Density 0.778 g/mL
Boiling Point 32°C (15 mmHg)
Quantity 2 g
UN Number UN1993
Formula Weight 138.28
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methyl-4-trimethylsilyl-1-buten-3-yne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.