missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(4-methylphenyl)-3-furoic acid, 97%, Thermo Scientific™

Product Code. 10504763
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10504763 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10504763 Supplier Thermo Scientific Maybridge Supplier No. MO01311DA
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 111787-86-1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C13H12O3
MDL Number MFCD05664429
Quantity 1 g
Synonym 2-methyl-5-4-methylphenyl-3-furoic acid, 2-methyl-5-4-methylphenyl furan-3-carboxylic acid, 2-methyl-5-p-tolyl furan-3-carboxylic acid, 3-furancarboxylic acid,2-methyl-5-4-methylphenyl, acmc-20mesz
InChI Key WWJLHTFFOPYRMV-UHFFFAOYSA-N
SMILES CC1=C(C=C(O1)C1=CC=C(C)C=C1)C(O)=O
IUPAC Name 2-methyl-5-(4-methylphenyl)furan-3-carboxylic acid
Molecular Weight (g/mol) 216.24
PubChem CID 2794742
Formula Weight 216.23
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-(4-methylphenyl)-3-furoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.