missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(methylthio)furan, 99%

Product Code. 11469477
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469477 10 g 10g
11459477 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469477 Supplier Thermo Scientific Alfa Aesar Supplier No. B21998.09
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13678-59-6 | C6H8OS | 128.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13678-59-6
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.189
MDL Number MFCD01208018
InChI Key RESBOJMQOGJOMW-UHFFFAOYSA-N
Synonym 2-methyl-5-methylthio furan, 5-methyl-2-methylthio furan, furan, 2-methyl-5-methylthio, methyl 5-methylfuryl sulfide, 2-methyl-5-methylsulfanyl furan, 2-methyl-5-methylthiofuran, unii-41z7b950jj, 2-methyl 5-methyl thio furan, furan,2-methyl-5-methylthio, fema no. 3366
PubChem CID 61657
IUPAC Name 2-methyl-5-methylsulfanylfuran
SMILES CC1=CC=C(O1)SC

Specifications

Density 1.063
Boiling Point 79°C to 81°C (50 mmHg)
Flash Point 50°C (122°F)
Odor Stench
Quantity 10 g
UN Number UN1993
Formula Weight 128.19
Percent Purity 99%
Chemical Name or Material 2-Methyl-5-(methylthio)furan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.