missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-phenylthiophene, 96%, Thermo Scientific™

Product Code. 11305679
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11305679 1 g 1g
11315679 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11305679 Supplier Thermo Scientific Alfa Aesar Supplier No. L20420.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Methyl-5-phenylthiophen is employed as intermediate for pharmaceuticals.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5069-26-1
Molecular Formula C11H10S
Molecular Weight (g/mol) 174.261
MDL Number MFCD06201035
InChI Key NEKXRPVOSZHILM-UHFFFAOYSA-N
Synonym acmc-1alda, 2-phenyl-5-methylthiophene, nekxrpvoszhilm-uhfffaoysa
PubChem CID 7578805
IUPAC Name 2-methyl-5-phenylthiophene
SMILES CC1=CC=C(S1)C2=CC=CC=C2

Specifications

Melting Point 46°C to 49°C
Odor Odorless
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 174.27
Percent Purity 96%
Chemical Name or Material 2-Methyl-5-phenylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.