missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-quinolinecarboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10055972
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10055972 250 mg 250mg
10371493 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10055972 Supplier Thermo Scientific Maybridge Supplier No. CC28401CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 635-80-3
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.198
MDL Number MFCD00748504
InChI Key IZONZQFTYGVOOO-UHFFFAOYSA-N
Synonym 2-methyl-6-quinolinecarboxylic acid, 6-quinolinecarboxylic acid, 2-methyl, 2-methyl-6-quinolinecarboxylicacid, 2-methylquinoline-6-carboxylicacid, acmc-1b4d8, quinaldine-6-carboxylic acid, 6-carboxy-2-methyl quinoline, ksc352o8f, 2-methyl-quinoline-6-carboxylic acid
PubChem CID 604483
IUPAC Name 2-methylquinoline-6-carboxylic acid
SMILES CC1=NC2=C(C=C1)C=C(C=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 2-Methyl-6-quinolinecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.