missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-methyl-benzoimidazol-1-yl)-acetic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
Specifications
Specifications
| CAS Min % | 97.0 |
| Assay Percent Range | 97% |
| Quantity | 250 mg |
| IUPAC Name | 2-(2-methyl-1H-1,3-benzodiazol-1-yl)acetate |
| Formula Weight | 190.2 |
| Percent Purity | 97% |
| Chemical Name or Material | (2-Methyl-benzoimidazol-1-yl)-acetic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction