missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-pentanoic Acid, TRC

Product Code. 30476784
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476784 50 g 50g
30482270 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30476784 Supplier Toronto Research Chemicals Supplier No. TRCM35855050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-61-0
Molecular Formula C6 H12 O2
Molecular Weight (g/mol) 116.16
Synonym (2RS)-2-Methylpentanoic acid, Pentanoic acid, 2-methyl-, Valeric acid, 2-methyl- (6CI,7CI,8CI), Valeric acid, α-methyl- (3CI), 2-Methylpentanoic acid, (±)-2-Methylpentanoic acid, 2-Methyl-n-valeric acid, 2-Methylvaleric acid, 2-Pentanecarboxylic acid, DL-2-Methylpentanoic acid, Methylpropylacetic acid, NSC 8406, α-Methylvaleric acid
IUPAC Name 2-methylpentanoic acid
SMILES CCCC(C)C(=O)O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H12O2/c1-3-4-5(2)6(7)8/h5H,3-4H2,1-2H3,(H,7,8)
Formula Weight 116.0837
Chemical Name or Material 2-Methylvaleric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.