missing translation for 'onlineSavingsMsg'
Learn More

2-Methylanthraquinone 94%, ACROS Organics™

Product Code. 10075803 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10075803 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10075803 Supplier Thermo Scientific Acros Supplier No. 149241000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 84-54-8
CAS Min % 92.0
CAS Max % 100.0
Melting Point >168°C
Color Green-Yellow to Yellow
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Molecular Formula C15H10O2
MDL Number MFCD00001235
Quantity 100 g
Beilstein 07, 809
Merck Index 15, 6095
Synonym 2-methylanthraquinone, tectoquinone, 2-methyl anthraquinone, techtoquinone, tectochinon, 2-methyl-9,10-anthraquinone, 9,10-anthracenedione, 2-methyl, beta-methylanthraquinone, 2-methylanthra-9,10-quinone, anthraquinone, 2-methyl
Solubility Information 2% max. Insoluble Matter (in chloroform)
InChI Key NJWGQARXZDRHCD-UHFFFAOYSA-N
SMILES CC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
IUPAC Name 2-methylanthracene-9,10-dione
Molecular Weight (g/mol) 222.243
PubChem CID 6773
ChEBI CHEBI:9427
Formula Weight 222.24
Percent Purity ≥92%
Physical Form Fine Powder
Chemical Name or Material 2-Methylanthraquinone, 94%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.