missing translation for 'onlineSavingsMsg'
Learn More

2-Methylanthraquinone, 97%

Product Code. 11439100
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439100 50 g 50g
11449100 250 g 250g
11429100 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439100 Supplier Thermo Scientific Alfa Aesar Supplier No. A14502.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 84-54-8 | C15H10O2 | 222.243 g/mol

2-Methylanthraquinone is used as a pharmaceutical intermediate. It is used in smog dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylanthraquinone is used as a pharmaceutical intermediate. It is used in smog dyes.

Solubility
Soluble in water 1.234 mg/L @ 25°C.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-54-8
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.243
MDL Number MFCD00001235
InChI Key NJWGQARXZDRHCD-UHFFFAOYSA-N
Synonym 2-methylanthraquinone, tectoquinone, 2-methyl anthraquinone, techtoquinone, tectochinon, 2-methyl-9,10-anthraquinone, 9,10-anthracenedione, 2-methyl, beta-methylanthraquinone, 2-methylanthra-9,10-quinone, anthraquinone, 2-methyl
PubChem CID 6773
ChEBI CHEBI:9427
IUPAC Name 2-methylanthracene-9,10-dione
SMILES CC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O

Specifications

Melting Point 170°C to 174°C
Flash Point 209°C (408°F)
Quantity 50 g
Beilstein 2050523
Merck Index 14,6021
Solubility Information Soluble in water 1.234mg/L @ 25°C.
Formula Weight 222.24
Percent Purity 97%
Chemical Name or Material 2-Methylanthraquinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.