missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzimidazole, 98%

Product Code. 10654982 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10654982 5 g Glass bottle 5g
10071410 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10654982 Supplier Thermo Scientific Acros Supplier No. 126330050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 615-15-6 | C8H8N2 | 132.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-15-6
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00005598
InChI Key LDZYRENCLPUXAX-UHFFFAOYSA-N
Synonym 2-methylbenzimidazole, 2-methyl-1h-benzo d imidazole, 1h-benzimidazole, 2-methyl, benzimidazole, 2-methyl, 2-methyl-1h-benzoimidazole, 2-methyl-1h-1,3-benzodiazole, acetamidine, n-n'-o-phenylene, 2-methyl benzimidazole, unii-zh8iww7y8b, 2-methylbenaimidazole
PubChem CID 11984
IUPAC Name 2-methyl-1H-benzimidazole
SMILES CC1=NC2=CC=CC=C2N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 175°C to 179°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 23, 145
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: soluble in ethanol,ether
Formula Weight 132.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylbenzimidazole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methylbenzimidazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.