missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzofuran, 96%, Thermo Scientific Chemicals

Catalog Number 11956201
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4265-25-2
C9H8O
132.16
MFCD00005850
GBGPVUAOTCNZPT-UHFFFAOYSA-N
2-methylbenzofuran, benzofuran, 2-methyl, 2-methylcumarone, 2-methylbenzo b furan, 2-methyl benzofuran, unii-8x3183bz3x, methylbenzofuran, methyl benzoxole, 2-methyl-benzofuran, #
20263
2-methyl-1-benzofuran
CC1=CC2=CC=CC=C2O1
This item is not returnable. View return policy
4265-25-2
C9H8O
132.16
MFCD00005850
GBGPVUAOTCNZPT-UHFFFAOYSA-N
2-methylbenzofuran, benzofuran, 2-methyl, 2-methylcumarone, 2-methylbenzo b furan, 2-methyl benzofuran, unii-8x3183bz3x, methylbenzofuran, methyl benzoxole, 2-methyl-benzofuran, #
20263
2-methyl-1-benzofuran
CC1=CC2=CC=CC=C2O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0570g/mL
Boiling Point 197.0°C to 198.0°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5618
Quantity 1 g
Beilstein 17,6
Specific Gravity 1.057
Formula Weight 132.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-Methylbenzofuran
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methylbenzofuran
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'